(S)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binaphthyl-2,2'-diyl-N-tosyl phosphoramide

Lieferant: Apollo Scientific

PC440010-100MG
APOSPC440010-100MGEA 582 EUR
APOSPC440010-100MG
(S)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binaphthyl-2,2'-diyl-N-tosyl phosphoramide
(S)-3,3'-Bis[3,5-bis(trifluoromethyl)phenyl]-1,1'-binaphthyl-2,2'-diyl-N-tosyl phosphoramide
Formel: C₄₃H₂₄F₁₂NO₅PS
Molecular Weight: 925,674 g/mol
Schmelzpunkt: 170…180 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28139336
CAS-Nummer: 1706437-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR