2-(4-Fluorophenyl)-3-nitronaphthalene

Lieferant: Apollo Scientific

PC440012-100MG
APOSPC440012-100MGEA 1760 EUR
APOSPC440012-100MG
2-(4-Fluorophenyl)-3-nitronaphthalene
2-(4-Fluorophenyl)-3-nitronaphthalene
Formel: C₁₆H₁₀FNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28154811
CAS-Nummer: 1450995-50-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR