1-(2-Amino-3-chloro-6-fluorophenyl)naphthalen-2-ol

Lieferant: Apollo Scientific

PC440014-50MG
APOSPC440014-50MGEA 1760 EUR
APOSPC440014-50MG
1-(2-Amino-3-chloro-6-fluorophenyl)naphthalen-2-ol
1-(2-Amino-3-chloro-6-fluorophenyl)naphthalen-2-ol
Formel: C₁₆H₁₁ClFNO
Molecular Weight: 287,716 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154813
CAS-Nummer: 1424386-52-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR