2-Fluoro-9H-carbazole

Lieferant: Apollo Scientific
Achtung

PC440015-100MG
APOSPC440015-100MGEA 535 EUR
APOSPC440015-100MG
2-Fluoro-9H-carbazole
2-Fluoro-9H-carbazole
Formel: C₁₂H₈FN
Molecular Weight: 185,20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09033508
CAS-Nummer: 391-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR