9-(Trifluoromethyl)-11H-benzo[a]carbazole

Lieferant: Apollo Scientific

PC440016-100MG
APOSPC440016-100MGEA 546 EUR
APOSPC440016-100MG
9-(Trifluoromethyl)-11H-benzo[a]carbazole
9-(Trifluoromethyl)-11H-benzo[a]carbazole
Formel: C₁₇H₁₀F₃N
Molecular Weight: 285,263 g/mol
Schmelzpunkt: 249…251 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD28154814
CAS-Nummer: 1607470-16-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR