9-Fluoro-11H-benzo[a]carbazole
Lieferant: Apollo Scientific
PC440017-100MG
APOSPC440017-100MGEA
546
EUR
APOSPC440017-100MG
9-Fluoro-11H-benzo[a]carbazole
9-Fluoro-11H-benzo[a]carbazole
Formel:
C₁₆H₁₀FN Lagertemperatur: Raumtemperatur |
MDL:
MFCD28154815 CAS-Nummer: 1041143-65-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...