2-(3-Fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one

Lieferant: Apollo Scientific

PC440018-100MG
APOSPC440018-100MGEA 626 EUR
APOSPC440018-100MG
2-(3-Fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one
2-(3-Fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one
Formel: C₁₆H₁₂FNO₃
Molecular Weight: 285,27 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154816
CAS-Nummer: 1451449-29-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR