2-(5-Chloro-2-fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one

Lieferant: Apollo Scientific

PC440019-100MG
APOSPC440019-100MGEA 546 EUR
APOSPC440019-100MG
2-(5-Chloro-2-fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one
2-(5-Chloro-2-fluoro-4-nitrophenyl)-3,4-dihydronaphthalen-1(2H)-one
Formel: C₁₆H₁₁ClFNO₃
Molecular Weight: 319,715 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154817
CAS-Nummer: 1451449-38-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR