1-(4-Fluorophenyl)-2-(4-nitrophenyl)ethan-1-one

Lieferant: Apollo Scientific

PC440020-100MG
APOSPC440020-100MGEA 626 EUR
APOSPC440020-100MG
1-(4-Fluorophenyl)-2-(4-nitrophenyl)ethan-1-one
1-(4-Fluorophenyl)-2-(4-nitrophenyl)ethan-1-one
Formel: C₁₄H₁₀FNO₃
Molecular Weight: 259,233 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16350559
CAS-Nummer: 140423-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR