2-(4-Fluorophenyl)benzofuran

Lieferant: Apollo Scientific
Achtung

PC440021-100MG
APOSPC440021-100MGEA 1470 EUR
APOSPC440021-100MG
2-(4-Fluorophenyl)benzofuran
2-(4-Fluorophenyl)benzofuran
Formel: C₁₄H₉FO
Molecular Weight: 212,219 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28154818
CAS-Nummer: 69976-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR