2-(4-(Trifluoromethyl)-phenyl)benzofuran

Lieferant: Apollo Scientific
Achtung

PC440022-100MG
APOSPC440022-100MGEA 1760 EUR
APOSPC440022-100MG
2-(4-(Trifluoromethyl)-phenyl)benzofuran
2-(4-(Trifluoromethyl)-phenyl)benzofuran
Formel: C₁₅H₉F₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD28154819
CAS-Nummer: 904325-87-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR