3-Fluoro-4-nitrophenylacetonitrile 95%

Lieferant: Apollo Scientific

PC4405-250MG
APOSPC4405-250MGEA 159 EUR
APOSPC4405-250MG
3-Fluoro-4-nitrophenylacetonitrile 95%
3-Fluoro-4-nitrophenylacetonitrile
Formel: C₈H₅FN₂O₂
Molecular Weight: 180,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03094238
CAS-Nummer: 503315-75-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR