3-Fluoro-4-nitrophenylacetic acid 95+%

Lieferant: Apollo Scientific

Synonyme: (3-Fluoro-4-nitrophenyl)acetic acid

PC4406-250MG
APOSPC4406-250MGEA 128 EUR
APOSPC4406-250MG
3-Fluoro-4-nitrophenylacetic acid 95+%
3-Fluoro-4-nitrophenylacetic acid
Formel: C₈H₆FNO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03094239
CAS-Nummer: 163395-24-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR