2-Fluoro-5-(methylsulphonyl)aniline 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC4413-1G
PC4413-5G
PC4413-25G
APOSPC4413-1GEA
65.4
EUR
APOSPC4413-1G
APOSPC4413-5G
APOSPC4413-25G
2-Fluoro-5-(methylsulphonyl)aniline 95+%
2-Fluoro-5-(methylsulphonyl)aniline
Formel:
C₇H₈FNO₂S Schmelzpunkt: 70…72 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01941314 CAS-Nummer: 387358-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...