Perfluorophenyl 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoate 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Morpholin-4-yl)-3-[(pentafluorophenoxy)carbonyl]benzotrifluoride, Pentafluorophenyl 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoate

PC4415-1G
APOSPC4415-1GEA 335 EUR
APOSPC4415-1G
Perfluorophenyl 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoate 97+%
Perfluorophenyl 2-(morpholin-4-yl)-5-(trifluoromethyl)benzoate
Formel: C₁₈H₁₁F₈NO₃
Molecular Weight: 441,272 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025890
CAS-Nummer: 898289-52-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR