5-Bromo-3-fluoro-2-hydroxypyridine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Bromo-3-fluoropyridin-2(1H)-one

PC445022-5G
APOSPC445022-5GEA 89.9 EUR
APOSPC445022-5G
5-Bromo-3-fluoro-2-hydroxypyridine 95%
5-Bromo-3-fluoro-2-hydroxypyridine
Formel: C₅H₃BrFNO
Molecular Weight: 191.986 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375017
CAS-Nummer: 156772-63-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR