2-Chlor-3-fluor-5-hydroxypyridin 97%

Lieferant: Apollo Scientific

PC445035-1G
APOSPC445035-1GEA 55.1 EUR
APOSPC445035-1G
2-Chlor-3-fluor-5-hydroxypyridin 97%
2-Chloro-3-fluoro-5-hydroxypyridine
Formel: C₆H₄ClFO
Molecular Weight: 147,5400 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03411573
CAS-Nummer: 870062-76-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR