3-Brom-2-chlor-5-fluorpyridin 95%

Lieferant: Apollo Scientific
Achtung

PC445039-25G
APOSPC445039-25GEA 108 EUR
APOSPC445039-25G
3-Brom-2-chlor-5-fluorpyridin 95%
3-Bromo-2-chloro-5-fluoropyridine
Formel: C₅H₂BrClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD03095269
CAS-Nummer: 884494-36-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR