6-Fluor-3-iod-2-methylpyridin 97%

Lieferant: Apollo Scientific

Synonyme: 6-Fluor-3-iod-2-picolin, 2-Fluor-5-iod-6-methylpyridin, 2-Fluor-5-iod-6-picolin

PC445047-1G
APOSPC445047-1GEA 56.7 EUR
APOSPC445047-1G
6-Fluor-3-iod-2-methylpyridin 97%
6-Fluor-3-iod-2-methylpyridin
Formel: C₆H₅FIN
Molecular Weight: 237,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08277264
CAS-Nummer: 884495-23-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR