5-Bromo-2-fluoro-3-nitropyridine 97%
Lieferant: Apollo Scientific
PC445050-1G
APOSPC445050-1GEA
60.9
EUR
APOSPC445050-1G
5-Bromo-2-fluoro-3-nitropyridine 97%
5-Bromo-2-fluoro-3-nitropyridine
Formel:
C₅H₂BrFN₂O₂ Molecular Weight: 220,984 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07375020 CAS-Nummer: 886372-98-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...