2-Bromo-5-fluoro-4-methylpyridine 98%

Lieferant: Apollo Scientific

PC445054-5G
APOSPC445054-5GEA 79.9 EUR
APOSPC445054-5G
2-Bromo-5-fluoro-4-methylpyridine 98%
2-Bromo-5-fluoro-4-methylpyridine
Formel: C₆H₅BrFN
Lagertemperatur: Raumtemperatur
MDL: MFCD08277310
CAS-Nummer: 885168-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR