5-Fluoro-4-iodo-2-methoxypyridine
Lieferant: Apollo Scientific
PC445060-1G
APOSPC445060-1GEA
173
EUR
APOSPC445060-1G
5-Fluoro-4-iodo-2-methoxypyridine
5-Fluoro-4-iodo-2-methoxypyridine
Formel:
C₆H₅FINO Molecular Weight: 253,013 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659503 CAS-Nummer: 884495-13-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...