4-Fluor-2-methoxybenzylamin 95+%
Lieferant: Apollo Scientific
Synonyme:
(4-Fluoro-2-methoxyphenyl)methanamine, (4-Fluoro- 2-methoxy-phenyl) methanamine
PC445061-1G
APOSPC445061-1GEA
120
EUR
APOSPC445061-1G
4-Fluor-2-methoxybenzylamin 95+%
4-Fluor-2-methoxybenzylamin
Formel:
C₈H₁₀FNO Molecular Weight: 155,17 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116361 CAS-Nummer: 870563-60-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...