Ammonium 2-perfluoropentoxy-2,3,3,3-tetrafluoropropanoate

Lieferant: Apollo Scientific
Achtung

PC4451-100G PC4451-25G PC4451-5G
APOSPC4451-100GEA 1040 EUR
APOSPC4451-100G APOSPC4451-25G APOSPC4451-5G
Ammonium 2-perfluoropentoxy-2,3,3,3-tetrafluoropropanoate
Ammonium 2-perfluoropentoxy-2,3,3,3-tetrafluoropropanoate
Formel: C₈HF₁₅O₃·H₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD07783987
CAS-Nummer: 96513-97-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR