2-(5,6-Difluoro-1H-benzoimidazol-2-yl)ethylamine

Lieferant: Apollo Scientific

PC446007-1G PC446007-500MG
APOSPC446007-1GEA 412 EUR
APOSPC446007-1G APOSPC446007-500MG
2-(5,6-Difluoro-1H-benzoimidazol-2-yl)ethylamine
2-(5,6-Difluoro-1H-benzoimidazol-2-yl)ethylamine
Formel: C₉H₉F₂N₃
Molecular Weight: 197.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10568201
CAS-Nummer: 1053656-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR