3-Chloro-5-(trifluoromethyl)pyridine-2-carbaldehyde

Lieferant: Apollo Scientific
Achtung

PC446013-1G
APOSPC446013-1GEA 0 EUR
APOSPC446013-1G
3-Chloro-5-(trifluoromethyl)pyridine-2-carbaldehyde
3-Chloro-5-(trifluoromethyl)pyridine-2-carbaldehyde
Formel: C₇H₃ClF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00153103
CAS-Nummer: 175277-50-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR