5-Chloromethyl-2-(4-(trifluoromethyl)phenyl)-1H-imidazole

Lieferant: Apollo Scientific

PC446024-1G PC446024-5G
APOSPC446024-1GEA 219 EUR
APOSPC446024-1G APOSPC446024-5G
5-Chloromethyl-2-(4-(trifluoromethyl)phenyl)-1H-imidazole
5-Chloromethyl-2-(4-(trifluoromethyl)phenyl)-1H-imidazole
Formel: C₁₁H₈ClF₃N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD10568333
CAS-Nummer: 466664-94-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR