C-[3-(5'-(Trifluoromethyl)pyridin-2'-yl)[1,2,4]oxadiazol-5-yl]methylammonium trifluoroacetate
Lieferant: Apollo Scientific
Synonyme:
1-{3-[5-(Trifluoromethyl)pyridin-2-yl]-1,2,4-oxadiazol-5-yl}methanamine trifluoroacetate
PC446027-1G
APOSPC446027-1GEA
528
EUR
APOSPC446027-1G
C-[3-(5'-(Trifluoromethyl)pyridin-2'-yl)[1,2,4]oxadiazol-5-yl]methylammonium trifluoroacetate
C-[3-(5'-(Trifluoromethyl)pyridin-2'-yl)[1,2,4]oxadiazol-5-yl]methylammonium trifluoroacetate
Formel:
C₁₁H₈F₆N₄O₃ Molecular Weight: 358,2 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12131103 CAS-Nummer: 1208081-75-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...