2-Chloromethyl-6-trifluoromethyl-1H-benzoimidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Chloromethyl)-6-(trifluoromethyl)-1H-benzo[d]imidazole
PC446030-1G
APOSPC446030-1GEA
217
EUR
APOSPC446030-1G
2-Chloromethyl-6-trifluoromethyl-1H-benzoimidazole
2-Chloromethyl-6-trifluoromethyl-1H-benzoimidazole
Formel:
C₉H₆ClF₃N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD08234908 CAS-Nummer: 107430-29-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...