2-Chloromethyl-6-fluoro-1H-benzoimidazole

Lieferant: Apollo Scientific

PC446031-1G PC446031-5G
APOSPC446031-1GEA 168 EUR
APOSPC446031-1G APOSPC446031-5G
2-Chloromethyl-6-fluoro-1H-benzoimidazole
2-Chloromethyl-6-fluoro-1H-benzoimidazole
Formel: C₈H₆ClFN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08234909
CAS-Nummer: 156144-42-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR