4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid tert-butyl ester

Lieferant: Apollo Scientific

PC446039-100MG
APOSPC446039-100MGEA 201 EUR
APOSPC446039-100MG
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid tert-butyl ester
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid tert-butyl ester
Formel: C₁₆H₂₁ClF₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19981199
CAS-Nummer: 1311278-11-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR