(4-Chloro-phenyl)-[4-(3-chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazin-1-yl]-methanone

Lieferant: Apollo Scientific

PC446040-100MG
APOSPC446040-100MGEA 201 EUR
APOSPC446040-100MG
(4-Chloro-phenyl)-[4-(3-chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazin-1-yl]-methanone
(4-Chloro-phenyl)-[4-(3-chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazin-1-yl]-methanone
Formel: C₁₈H₁₆Cl₂F₃N₃O
Molecular Weight: 418,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19981201
CAS-Nummer: 1311278-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR