4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid (4-chloro-phenyl)-amide

Lieferant: Apollo Scientific

PC446041-100MG
APOSPC446041-100MGEA 201 EUR
APOSPC446041-100MG
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid (4-chloro-phenyl)-amide
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carboxylic acid (4-chloro-phenyl)-amide
Formel: C₁₈H₁₇Cl₂F₃N₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD19981204
CAS-Nummer: 1311279-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR