4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carbothioic acid (4-chloro-phenyl)-amide

Lieferant: Apollo Scientific

PC446042-100MG
APOSPC446042-100MGEA 201 EUR
APOSPC446042-100MG
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carbothioic acid (4-chloro-phenyl)-amide
4-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-piperazine-1-carbothioic acid (4-chloro-phenyl)-amide
Formel: C₁₈H₁₇Cl₂F₃N₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD19981205
CAS-Nummer: 1311278-34-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR