Dimethyl{6-[4-(methylhydrazonomethyl)phenyl]-4-trifluoromethylpyridin-2-yl}amine

Lieferant: Apollo Scientific

PC446047-1G
APOSPC446047-1GEA 474 EUR
APOSPC446047-1G
Dimethyl{6-[4-(methylhydrazonomethyl)phenyl]-4-trifluoromethylpyridin-2-yl}amine
Dimethyl{6-[4-(methylhydrazonomethyl)phenyl]-4-trifluoromethylpyridin-2-yl}amine
Formel: C₁₆H₁₇F₃N₄
Lagertemperatur: Raumtemperatur
MDL: MFCD18384856
CAS-Nummer: 1300019-70-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR