4-Fluoro-3-prop-2-ynyloxybenzoic acid methylester

Lieferant: Apollo Scientific

PC446048-1G
APOSPC446048-1GEA 201 EUR
APOSPC446048-1G
4-Fluoro-3-prop-2-ynyloxybenzoic acid methylester
4-Fluoro-3-prop-2-ynyloxybenzoic acid methylester
Formel: C₁₁H₉FO₃
Molecular Weight: 208,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15142794
CAS-Nummer: 214822-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR