[Methyl(2-trifluoromethylquinazolin-4-yl)amino]acetic acid ethyl ester

Lieferant: Apollo Scientific

PC446049-1G
APOSPC446049-1GEA 265 EUR
APOSPC446049-1G
[Methyl(2-trifluoromethylquinazolin-4-yl)amino]acetic acid ethyl ester
[Methyl(2-trifluoromethylquinazolin-4-yl)amino]acetic acid ethyl ester
Formel: C₁₄H₁₄F₃N₃O₂
Molecular Weight: 313,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15142795
CAS-Nummer: 1208081-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR