[Methyl(2-trifluoromethylquinazolin-4-yl)amino]acetic acid

Lieferant: Apollo Scientific

PC446050-1G
APOSPC446050-1GEA 176 EUR
APOSPC446050-1G
[Methyl(2-trifluoromethylquinazolin-4-yl)amino]acetic acid
[Methyl(2-trifluoromethylquinazolin-4-yl)amino]acetic acid
Formel: C₁₂H₁₀F₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD08758433
CAS-Nummer: 927969-23-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR