[3-(4-Aminophenyl)-5-trifluoromethylpyridin-2-yl]dimethylamine

Lieferant: Apollo Scientific

PC446052-1G
APOSPC446052-1GEA 391 EUR
APOSPC446052-1G
[3-(4-Aminophenyl)-5-trifluoromethylpyridin-2-yl]dimethylamine
[3-(4-Aminophenyl)-5-trifluoromethylpyridin-2-yl]dimethylamine
Formel: C₁₄H₁₄F₃N₃
Molecular Weight: 281,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384837
CAS-Nummer: 1299607-52-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR