1-[4-(6-Dimethylamino-4-trifluoromethyl-pyridin-2-yl)-phenyl]-ethanone
Lieferant: Apollo Scientific
PC446057-500MG
APOSPC446057-500MGEA
265
EUR
APOSPC446057-500MG
1-[4-(6-Dimethylamino-4-trifluoromethyl-pyridin-2-yl)-phenyl]-ethanone
1-[4-(6-Dimethylamino-4-trifluoromethyl-pyridin-2-yl)-phenyl]-ethanone
Formel:
C₁₆H₁₅F₃N₂O Molecular Weight: 308,298 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19981356 CAS-Nummer: 1311280-01-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...