6-Fluoro-quinoxaline-2,3-diol

Lieferant: Apollo Scientific

PC446058-1G PC446058-500MG
APOSPC446058-1GEA 317 EUR
APOSPC446058-1G APOSPC446058-500MG
6-Fluoro-quinoxaline-2,3-diol
6-Fluoro-quinoxaline-2,3-diol
Formel: C₈H₅FN₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00629759
CAS-Nummer: 61875-34-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR