2-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-5-methyl-2H-pyrazole-3-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

PC446062-100MG
APOSPC446062-100MGEA 201 EUR
APOSPC446062-100MG
2-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-5-methyl-2H-pyrazole-3-carboxylic acid ethyl ester
2-(3-Chloro-5-trifluoromethyl-pyridin-2-ylmethyl)-5-methyl-2H-pyrazole-3-carboxylic acid ethyl ester
Formel: C₁₄H₁₃ClF₃N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD20526237
CAS-Nummer: 1351479-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR