4-(6-Dimethylamino-4-trifluoromethylpyridin-2-yl)benzoic acid methyl ester

Lieferant: Apollo Scientific

PC446063-1G
APOSPC446063-1GEA 350 EUR
APOSPC446063-1G
4-(6-Dimethylamino-4-trifluoromethylpyridin-2-yl)benzoic acid methyl ester
4-(6-Dimethylamino-4-trifluoromethylpyridin-2-yl)benzoic acid methyl ester
Formel: C₁₆H₁₅F₃N₂O₂
Molecular Weight: 324.31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384852
CAS-Nummer: 1299607-60-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR