[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester

Lieferant: Apollo Scientific

PC446064-1G PC446064-5G
APOSPC446064-1GEA 244 EUR
APOSPC446064-1G APOSPC446064-5G
[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester
[3-Chloro-5-trifluoromethyl-1H-pyridin-(2E)-ylidene]cyanoacetic acid ethyl ester
Formel: C₁₁H₈ClF₃N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18384848
CAS-Nummer: 1300019-72-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR