1-Phenyl-3-trifluoromethyl-1H-pyrazole-4-carboxylic acid

Lieferant: Apollo Scientific

PC446065-1G
APOSPC446065-1GEA 201 EUR
APOSPC446065-1G
1-Phenyl-3-trifluoromethyl-1H-pyrazole-4-carboxylic acid
1-Phenyl-3-trifluoromethyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₁H₇F₃N₂O₂
Molecular Weight: 256,181 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11707231
CAS-Nummer: 142818-01-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR