3-Nitro-5-trifluoromethyl-benzoic acid ethyl ester
Lieferant: Apollo Scientific
PC446067-1G
APOSPC446067-1GEA
321
EUR
APOSPC446067-1G
3-Nitro-5-trifluoromethyl-benzoic acid ethyl ester
3-Nitro-5-trifluoromethyl-benzoic acid ethyl ester
Formel:
C₁₀H₈F₃NO₄ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12025280 CAS-Nummer: 203513-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...