Methyl-1-(4-fluorobenzyl)-5-oxopyrrolidine-3-carboxylate

Lieferant: Apollo Scientific

Synonyme: 1-(4-Fluoro-benzyl)-5-oxo-pyrrolidine-3-carboxylic acid methyl ester

PC446068-1G
APOSPC446068-1GEA 244 EUR
APOSPC446068-1G
Methyl-1-(4-fluorobenzyl)-5-oxopyrrolidine-3-carboxylate
Methyl-1-(4-fluorobenzyl)-5-oxopyrrolidine-3-carboxylate
Formel: C₁₃H₁₄FNO₃
Molecular Weight: 251.26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13250867
CAS-Nummer: 1171317-52-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR