(5-Trifluoromethyl-pyridine-2-sulphonylamino)-acetic acid ethyl ester

Lieferant: Apollo Scientific

PC446069-1G
APOSPC446069-1GEA 412 EUR
APOSPC446069-1G
(5-Trifluoromethyl-pyridine-2-sulphonylamino)-acetic acid ethyl ester
(5-Trifluoromethyl-pyridine-2-sulphonylamino)-acetic acid ethyl ester
Formel: C₁₀H₁₁F₃N₂O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD21362494
CAS-Nummer: 1355334-83-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR