4-(5-Bromo-3-trifluoromethyl-pyridin-2-yl)-piperazine-1-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

PC446071-1G
APOSPC446071-1GEA 350 EUR
APOSPC446071-1G
4-(5-Bromo-3-trifluoromethyl-pyridin-2-yl)-piperazine-1-carboxylic acid ethyl ester
4-(5-Bromo-3-trifluoromethyl-pyridin-2-yl)-piperazine-1-carboxylic acid ethyl ester
Formel: C₁₃H₁₅BrF₃N₃O₂
Molecular Weight: 382.176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22573503
CAS-Nummer: 1401522-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR