1-(3-Trifluoromethyl-benzyl)-1H-pyrazole-4-carboxylic acid ethyl ester

Lieferant: Apollo Scientific

PC446073-1G
APOSPC446073-1GEA 402 EUR
APOSPC446073-1G
1-(3-Trifluoromethyl-benzyl)-1H-pyrazole-4-carboxylic acid ethyl ester
1-(3-Trifluoromethyl-benzyl)-1H-pyrazole-4-carboxylic acid ethyl ester
Formel: C₁₄H₁₃F₃N₂O₂
Molecular Weight: 298,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24386141
CAS-Nummer: 1035224-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR